1-(3-Chloro-2-fluorophenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

160420-5G 160420-25G 160420-1G 160420-500MG
77631-380EA 1484.23 USD
77631-380 77653-684 77640-754 77644-714
1-(3-Chloro-2-fluorophenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(3-Chloro-2-fluorophenyl)-2,2-dimethylpropan-1-ol

Precaución: For Research Use Only

Formula: C₁₁H₁₄ClFO
MW: 216.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20401007
CAS Number: 1339815-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR