4-(1-Methyl-4-(trifluoromethyl)-1H-imidazol-2-yl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

159366-5G 159366-25G 159366-1G 159366-500MG
77634-092EA 327 USD
77634-092 77653-680 77640-752 77648-794
4-(1-Methyl-4-(trifluoromethyl)-1H-imidazol-2-yl)benzonitrile ≥97%
4-(1-Methyl-4-(trifluoromethyl)-1H-imidazol-2-yl)benzonitrile

Precaución: For Research Use Only

Formula: C₁₂H₈F₃N₃
Storage Temperature: Ambient
MDL Number: MFCD34181775
CAS Number: 98298-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR