4-Fluoro-2-(methoxymethoxy)-1-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

15916-5G 15916-1G 15916-25G 15916-500MG
77634-090EA 302 USD
77634-090 77636-738 77653-678 77644-712
4-Fluoro-2-(methoxymethoxy)-1-methylbenzene ≥95%
4-Fluoro-2-(methoxymethoxy)-1-methylbenzene

Precaución: For Research Use Only

Formula: 170.181 g/mol
MW: C₉H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD31376248
CAS Number: 1067225-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR