2-Fluoro-6-(pyrrolidin-1-yl)pyridine ≥95%

Supplier: AOBChem USA

Warning

15855-5G 15855-1G 15855-500MG 15855-25G
77634-088EA 451 USD
77634-088 77636-734 77652-366 77657-536
2-Fluoro-6-(pyrrolidin-1-yl)pyridine ≥95%
2-Fluoro-6-(pyrrolidin-1-yl)pyridine

Precaución: For Research Use Only

Formula: C₉H₁₁FN₂
MW: 166.195 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10699460
CAS Number: 1000981-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR