1-(Benzo[d][1,3]dioxol-5-yl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

158462-5G 158462-1G 158462-500MG
77657-532EA 2083 USD
77657-532 77640-750 77644-708
1-(Benzo[d][1,3]dioxol-5-yl)cyclobutanol ≥95%
1-(Benzo[d][1,3]dioxol-5-yl)cyclobutanol

Precaución: For Research Use Only

Formula: 192.211 g/mol
MW: C₁₁H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34846760
CAS Number: 1521393-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR