5-Bromo-4-methyl-2-(trifluoromethyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

15837-5G 15837-25G 15837-1G 15837-500MG
77634-086EA 515 USD
77634-086 77653-674 77640-748 77644-706
5-Bromo-4-methyl-2-(trifluoromethyl)pyridine ≥95%
5-Bromo-4-methyl-2-(trifluoromethyl)pyridine

Precaución: For Research Use Only

Formula: C₇H₅BrF₃N
MW: 240.021 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257679
CAS Number: 1010422-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR