2-Chloro-5-(trifluoromethyl)pyridine-3-boronic acid ≥95%

Supplier: AOBChem USA

Warning

15817-5G 15817-1G 15817-25G
77634-080EA 198 USD
77634-080 77636-726 77653-672
2-Chloro-5-(trifluoromethyl)pyridine-3-boronic acid ≥95%
2-Chloro-5-(trifluoromethyl)pyridine-3-boronic acid

Precaución: For Research Use Only

Formula: C₆H₄BClF₃NO₂
MW: 225.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10696241
CAS Number: 536693-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR