Methyl 4,6-dichloro-5-fluoronicotinate ≥97%

Supplier: AOBChem USA

Warning

158051-1G 158051-500MG
77634-078EA 1499 USD
77634-078 77648-784
Methyl 4,6-dichloro-5-fluoronicotinate ≥97%
Methyl 4,6-dichloro-5-fluoronicotinate

Precaución: For Research Use Only

Formula: 224.017 g/mol
MW: C₇H₄Cl₂FNO₂
Storage Temperature: Ambient
MDL Number: MFCD28666394
CAS Number: 1588977-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR