2,2,2-Trifluoro-1-(4-(tert-pentyl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

158003-5G 158003-25G 158003-1G 158003-500MG
77631-696EA 1381 USD
77631-696 77653-670 77640-744 77644-702
2,2,2-Trifluoro-1-(4-(tert-pentyl)phenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(4-(tert-pentyl)phenyl)ethanone

Precaución: For Research Use Only

Formula: 244.253 g/mol
MW: C₁₃H₁₅F₃O
Storage Temperature: Ambient
MDL Number: MFCD07380783
CAS Number: 1505209-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR