1,3-Dichloro-4-(cyclopropylmethoxy)-2-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

157664-5G 157664-1G 157664-25G 157664-500MG
77631-370EA 1484.23 USD
77631-370 77636-724 77653-666 77644-698
1,3-Dichloro-4-(cyclopropylmethoxy)-2-iodobenzene ≥95%
1,3-Dichloro-4-(cyclopropylmethoxy)-2-iodobenzene

Precaución: For Research Use Only

Formula: C₁₀H₉Cl₂IO
Storage Temperature: Ambient
MDL Number: MFCD34846327
CAS Number: 2921751-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR