(2,6-Dichloro-3-(cyclopropylmethoxy)phenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

157663-5G 157663-25G 157663-1G 157663-500MG
77631-692EA 1441 USD
77631-692 77657-520 77640-738 77644-696
(2,6-Dichloro-3-(cyclopropylmethoxy)phenyl)(methyl)sulfane ≥95%
(2,6-Dichloro-3-(cyclopropylmethoxy)phenyl)(methyl)sulfane

Precaución: For Research Use Only

Formula: 263.183 g/mol
MW: C₁₁H₁₂Cl₂OS
Storage Temperature: Ambient
MDL Number: MFCD34846326
CAS Number: 2921841-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR