1-Bromo-5-fluoro-2-iodo-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

15728-5G 15728-1G 15728-25G 15728-500MG
77631-688EA 856 USD
77631-688 77636-718 77653-662 77644-694
1-Bromo-5-fluoro-2-iodo-4-methoxybenzene ≥97%
1-Bromo-5-fluoro-2-iodo-4-methoxybenzene

Precaución: For Research Use Only

Formula: 330.921 g/mol
MW: C₇H₅BrFIO
Storage Temperature: Ambient
MDL Number: MFCD27937163
CAS Number: 1823954-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR