(2-Fluoro-6-methoxy-4-(trifluoromethyl)phenyl)(methyl)sulfane ≥95%
Supplier: AOBChem USA
|
Warning
|
157163-5G
157163-25G
157163-1G
157163-500MG
77631-686EA
1441
USD
77631-686
77653-660
77640-734
77648-776
(2-Fluoro-6-methoxy-4-(trifluoromethyl)phenyl)(methyl)sulfane ≥95%
(2-Fluoro-6-methoxy-4-(trifluoromethyl)phenyl)(methyl)sulfane
Precaución: For Research Use Only
Formula:
C₉H₈F₄OS Storage Temperature: Ambient |
MDL Number:
MFCD34845926 CAS Number: 2706787-04-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...