2-Fluoro-6-methoxy-4-(trifluoromethyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

157160-5G 157160-25G 157160-1G 157160-500MG
77631-366EA 1709.8 USD
77631-366 77653-658 77640-732 77644-692
2-Fluoro-6-methoxy-4-(trifluoromethyl)benzoic acid ≥95%
2-Fluoro-6-methoxy-4-(trifluoromethyl)benzoic acid

Precaución: For Research Use Only

Formula: 238.136 g/mol
MW: C₉H₆F₄O₃
Storage Temperature: Ambient
MDL Number: MFCD27975464
CAS Number: 1823863-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR