1-Bromo-2-chloro-4-(methoxymethoxy)-benzene ≥95%

Supplier: AOBChem USA

Warning

15710-5G 15710-1G 15710-25G 15710-500MG
77634-076EA 536 USD
77634-076 77636-716 77653-654 77648-774
1-Bromo-2-chloro-4-(methoxymethoxy)-benzene ≥95%
1-Bromo-2-chloro-4-(methoxymethoxy)-benzene

Precaución: For Research Use Only

Formula: 251.505 g/mol
MW: C₈H₈BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD31619368
CAS Number: 1300743-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR