6-Chloro-2-fluoro-3-hydroxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

15626-5G 15626-1G 15626-500MG
77631-364EA 1727.31 USD
77631-364 77636-714 77648-768
6-Chloro-2-fluoro-3-hydroxybenzoic acid ≥97%
6-Chloro-2-fluoro-3-hydroxybenzoic acid

Precaución: For Research Use Only

Formula: C₇H₄ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD10566791
CAS Number: 91659-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR