2-Fluoro-1-iodo-4-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-4-iodophenyl trifluoromethyl ether

156166-5G 156166-25G
77634-074EA 177 USD
77634-074 77653-646
2-Fluoro-1-iodo-4-(trifluoromethoxy)benzene ≥97%
2-Fluoro-1-iodo-4-(trifluoromethoxy)benzene

Precaución: For Research Use Only

Formula: C₇H₃F₄IO
MW: 305.996 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20526760
CAS Number: 1321963-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR