1-Chloro-2-fluoro-4-(trifluoromethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

156165-1G 156165-5G 156165-500MG
77636-712EA 628 USD
77636-712 77653-644 77644-686
1-Chloro-2-fluoro-4-(trifluoromethoxy)benzene ≥95%
1-Chloro-2-fluoro-4-(trifluoromethoxy)benzene

Precaución: For Research Use Only

Formula: 214.545 g/mol
MW: C₇H₃ClF₄O
Storage Temperature: Ambient
MDL Number: MFCD11036239
CAS Number: 1404194-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR