6-Bromo-2-fluoro-3-propoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15616-5G 15616-1G 15616-25G 15616-500MG
77634-072EA 288 USD
77634-072 77636-710 77653-642 77648-762
6-Bromo-2-fluoro-3-propoxybenzaldehyde ≥97%
6-Bromo-2-fluoro-3-propoxybenzaldehyde

Precaución: For Research Use Only

Formula: 261.088 g/mol
MW: C₁₀H₁₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD11110259
CAS Number: 1114809-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR