6-Bromo-3-ethoxy-2-fluorobenzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
15611-5G
15611-1G
15611-25G
15611-500MG
77634-070EA
366
USD
77634-070
77636-708
77657-516
77648-760
6-Bromo-3-ethoxy-2-fluorobenzaldehyde ≥97%
6-Bromo-3-ethoxy-2-fluorobenzaldehyde
Precaución: For Research Use Only
Formula:
C₉H₈BrFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD11110258 CAS Number: 1114809-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...