2-(Benzyloxy)-6-fluorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

15597-5G 15597-1G 15597-500MG
77631-362EA 1484.23 USD
77631-362 77636-706 77644-684
2-(Benzyloxy)-6-fluorobenzaldehyde ≥95%
2-(Benzyloxy)-6-fluorobenzaldehyde

Precaución: For Research Use Only

Formula: 230.234 g/mol
MW: C₁₄H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD12031619
CAS Number: 701265-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR