1-(Benzyloxy)-3-chlorobenzene ≥97%

Supplier: AOBChem USA

Warning

15596-5G 15596-25G 15596-1G 15596-500MG
77634-068EA 529 USD
77634-068 77657-514 77640-722 77648-758
1-(Benzyloxy)-3-chlorobenzene ≥97%
1-(Benzyloxy)-3-chlorobenzene

Precaución: For Research Use Only

Formula: C₁₃H₁₁ClO
MW: 218.67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05863215
CAS Number: 24318-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR