Tert-butyl 4-(4-chlorophenyl)-4-hydroxypiperidine-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

155806-5G 155806-1G 155806-25G 155806-500MG
77634-066EA 529 USD
77634-066 77636-704 77653-640 77648-756
Tert-butyl 4-(4-chlorophenyl)-4-hydroxypiperidine-1-carboxylate ≥95%
Tert-butyl 4-(4-chlorophenyl)-4-hydroxypiperidine-1-carboxylate

Precaución: For Research Use Only

Formula: 311.804 g/mol
MW: C₁₆H₂₂ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD09953408
CAS Number: 235109-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR