1-Bromo-4-cyclopentyl-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

154833-5G 154833-1G 154833-25G 154833-500MG
77631-360EA 1703.62 USD
77631-360 77636-702 77653-638 77644-676
1-Bromo-4-cyclopentyl-2-methylbenzene ≥95%
1-Bromo-4-cyclopentyl-2-methylbenzene

Precaución: For Research Use Only

Formula: 239.1515 g/mol
MW: C₁₂H₁₅Br
Storage Temperature: Ambient
MDL Number: MFCD34844442
CAS Number: 2969418-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR