3,6-Difluorobenzene-1,2-diol ≥97%

Supplier: AOBChem USA

Warning

154818-5G 154818-1G 154818-500MG
77653-636EA 2233 USD
77653-636 77640-718 77648-754
3,6-Difluorobenzene-1,2-diol ≥97%
3,6-Difluorobenzene-1,2-diol

Precaución: For Research Use Only

Formula: 146.092 g/mol
MW: C₆H₄F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18394833
CAS Number: 75626-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR