2-Bromo-1-(difluoromethoxy)-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

154518-5G 154518-25G 154518-1G 154518-500MG
77631-674EA 862 USD
77631-674 77653-630 77640-714 77648-748
2-Bromo-1-(difluoromethoxy)-4-(trifluoromethyl)benzene ≥97%
2-Bromo-1-(difluoromethoxy)-4-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: C₈H₄BrF₅O
MW: 291.013 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11036074
CAS Number: 954236-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR