5-Hydroxy-2-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

154220-5G 154220-25G 154220-1G 154220-500MG
77631-672EA 1357 USD
77631-672 77653-628 77640-712 77644-674
5-Hydroxy-2-(trifluoromethyl)benzaldehyde ≥97%
5-Hydroxy-2-(trifluoromethyl)benzaldehyde

Precaución: For Research Use Only

Formula: C₈H₅F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD16999729
CAS Number: 1243458-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR