2-Ethyl-1,3-difluoro-4-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

153828-5G 153828-25G 153828-1G 153828-500MG
77631-356EA 1599.59 USD
77631-356 77653-626 77640-710 77648-746
2-Ethyl-1,3-difluoro-4-methoxybenzene ≥95%
2-Ethyl-1,3-difluoro-4-methoxybenzene

Precaución: For Research Use Only

Formula: 172.172 g/mol
MW: C₉H₁₀F₂O
Storage Temperature: Ambient
MDL Number: MFCD34843721
CAS Number: 2710631-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR