2-Chloro-6-fluoro-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15326-5G 15326-1G 15326-500MG
77631-352EA 1599.59 USD
77631-352 77636-696 77644-664
2-Chloro-6-fluoro-4-methylbenzaldehyde ≥97%
2-Chloro-6-fluoro-4-methylbenzaldehyde

Precaución: For Research Use Only

Formula: C₈H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD28135628
CAS Number: 1534389-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR