2,5-Dibromo-6-fluorobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3,6-Dibromo-2-fluorobenzonitrile

152460-1G 152460-5G 152460-500MG
77634-060EA 785 USD
77634-060 77657-504 77648-740
2,5-Dibromo-6-fluorobenzonitrile ≥97%
2,5-Dibromo-6-fluorobenzonitrile

Precaución: For Research Use Only

Formula: C₇H₂Br₂FN
MW: 278.903 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846055
CAS Number: 1160574-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR