2-Benzyloxy-3-pyridineboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

15174-5G 15174-25G 15174-1G 15174-500MG
77634-058EA 456 USD
77634-058 77657-500 77640-704 77648-732
2-Benzyloxy-3-pyridineboronic acid pinacol ester ≥95%
2-Benzyloxy-3-pyridineboronic acid pinacol ester

Precaución: For Research Use Only

Formula: C₁₈H₂₂BNO₃
Storage Temperature: Ambient
MDL Number: MFCD06798265
CAS Number: 1073371-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR