6-Bromo-2-Chloro-3-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

15170-5G 15170-1G 15170-25G
77634-056EA 199 USD
77634-056 77636-688 77653-620
6-Bromo-2-Chloro-3-methylbenzaldehyde ≥95%
6-Bromo-2-Chloro-3-methylbenzaldehyde

Precaución: For Research Use Only

Formula: C₈H₆BrClO
Storage Temperature: Ambient
MDL Number: MFCD11040411
CAS Number: 1114808-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR