2,3-Dibromo-benzoic acid methyl ester ≥97%

Supplier: AOBChem USA

Warning

15167-5G 15167-1G 15167-25G 15167-500MG
77634-054EA 519 USD
77634-054 77636-686 77653-618 77648-730
2,3-Dibromo-benzoic acid methyl ester ≥97%
2,3-Dibromo-benzoic acid methyl ester

Precaución: For Research Use Only

Formula: C₈H₆Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD08752423
CAS Number: 881667-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR