2,3-Dibromobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

15164-5G 15164-25G 15164-1G 15164-500MG
77631-668EA 1369 USD
77631-668 77657-498 77640-702 77648-728
2,3-Dibromobenzonitrile ≥97%
2,3-Dibromobenzonitrile

Precaución: For Research Use Only

Formula: C₇H₃Br₂N
MW: 260.91 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08752371
CAS Number: 34362-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR