6-Bromo-3-chloro-2-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

151558-1G 151558-5G 151558-500MG
77636-684EA 574 USD
77636-684 77653-616 77644-658
6-Bromo-3-chloro-2-methoxyphenol ≥97%
6-Bromo-3-chloro-2-methoxyphenol

Precaución: For Research Use Only

Formula: C₇H₆BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD33418334
CAS Number: 1785582-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR