3,6-Dibromo-2-methoxyphenol ≥95%

Supplier: AOBChem USA

Warning

151557-1G 151557-5G 151557-500MG
77636-682EA 616 USD
77636-682 77657-496 77644-656
3,6-Dibromo-2-methoxyphenol ≥95%
3,6-Dibromo-2-methoxyphenol

Precaución: For Research Use Only

Formula: 281.929 g/mol
MW: C₇H₆Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34842255
CAS Number: 2091675-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR