4-Iodo-5-methoxybenzo[d][1,3]dioxole ≥95%

Supplier: AOBChem USA

Warning

151267-5G 151267-25G 151267-1G 151267-500MG
77631-346EA 1484.23 USD
77631-346 77635-460 77636-680 77644-654
4-Iodo-5-methoxybenzo[d][1,3]dioxole ≥95%
4-Iodo-5-methoxybenzo[d][1,3]dioxole

Precaución: For Research Use Only

Formula: 278.044 g/mol
MW: C₈H₇IO₃
Storage Temperature: Ambient
MDL Number: MFCD34842146
CAS Number: 2624384-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR