(5-Methoxybenzo[d][1,3]dioxol-4-yl)boronic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
151260-5G
151260-1G
151260-25G
151260-500MG
77631-344EA
1530.58
USD
77631-344
77636-676
77653-612
77644-650
(5-Methoxybenzo[d][1,3]dioxol-4-yl)boronic acid ≥95%
(5-Methoxybenzo[d][1,3]dioxol-4-yl)boronic acid
Precaución: For Research Use Only
Formula:
195.965 g/mol MW: C₈H₉BO₅ Storage Temperature: Ambient |
MDL Number:
MFCD30070649 CAS Number: 685516-54-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...