Methyl 4-cyano-2-fluoro-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

150548-1G 150548-5G 150548-500MG
77634-050EA 822 USD
77634-050 77657-490 77648-720
Methyl 4-cyano-2-fluoro-5-methylbenzoate ≥97%
Methyl 4-cyano-2-fluoro-5-methylbenzoate

Precaución: For Research Use Only

Formula: 193.174 g/mol
MW: C₁₀H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD23707560
CAS Number: 1427372-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR