2-Chlor-4-fluoro-5-(methoxycarbonyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

15018-5G 15018-25G 15018-1G 15018-500MG
77634-048EA 588 USD
77634-048 77653-602 77640-692 77644-646
2-Chlor-4-fluoro-5-(methoxycarbonyl)phenylboronic acid ≥95%
2-Chlor-4-fluoro-5-(methoxycarbonyl)phenylboronic acid

Precaución: For Research Use Only

Formula: C₈H₇BClFO₄
MW: 232.401 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878351
CAS Number: 957066-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR