1-Ethyl-4-fluoro-2-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

149244-5G 149244-25G 149244-1G 149244-500MG
77631-340EA 1797.35 USD
77631-340 77653-600 77640-690 77648-716
1-Ethyl-4-fluoro-2-iodobenzene ≥95%
1-Ethyl-4-fluoro-2-iodobenzene

Precaución: For Research Use Only

Formula: C₈H₈FI
Storage Temperature: Ambient
MDL Number: MFCD22490752
CAS Number: 1369791-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR