3-Bromo-4,5-dichlorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

149219-5G 149219-1G 149219-25G 149219-500MG
77634-046EA 568 USD
77634-046 77636-674 77657-488 77648-714
3-Bromo-4,5-dichlorobenzoic acid ≥97%
3-Bromo-4,5-dichlorobenzoic acid

Precaución: For Research Use Only

Formula: 269.9075 g/mol
MW: C₇H₃BrCl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD24107199
CAS Number: 791137-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR