(2-Isopropoxy-5-(trifluoromethyl)phenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

149030-5G 149030-1G 149030-25G 149030-500MG
77631-664EA 1441 USD
77631-664 77636-672 77653-598 77648-712
(2-Isopropoxy-5-(trifluoromethyl)phenyl)(methyl)sulfane ≥95%
(2-Isopropoxy-5-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: For Research Use Only

Formula: 250.281 g/mol
MW: C₁₁H₁₃F₃OS
Storage Temperature: Ambient
MDL Number: MFCD34840501
CAS Number: 2921874-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR