2-Isopropoxy-5-(trifluoromethyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

149028-5G 149028-1G 149028-500MG
77653-596EA 2140 USD
77653-596 77640-688 77644-644
2-Isopropoxy-5-(trifluoromethyl)benzaldehyde ≥95%
2-Isopropoxy-5-(trifluoromethyl)benzaldehyde

Precaución: For Research Use Only

Formula: 232.199 g/mol
MW: C₁₁H₁₁F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD29121495
CAS Number: 1289150-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR