2-Isopropoxy-5-(trifluoromethyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

149026-5G 149026-1G 149026-500MG 149026-25G
77631-338EA 1625.34 USD
77631-338 77636-670 77652-362 77657-486
2-Isopropoxy-5-(trifluoromethyl)benzoic acid ≥95%
2-Isopropoxy-5-(trifluoromethyl)benzoic acid

Precaución: For Research Use Only

Formula: C₁₁H₁₁F₃O₃
MW: 248.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17378712
CAS Number: 1273867-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR