2-Fluoro-4-iodo-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

14860-5G 14860-1G 14860-500MG 14860-25G
77631-662EA 884 USD
77631-662 77636-668 77652-360 77657-484
2-Fluoro-4-iodo-3-methoxybenzaldehyde ≥97%
2-Fluoro-4-iodo-3-methoxybenzaldehyde

Precaución: For Research Use Only

Formula: 280.035 g/mol
MW: C₈H₆FIO₂
Storage Temperature: Ambient
MDL Number: MFCD34564009
CAS Number: 861928-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR