2-(Cyclopropylmethoxy)-1,3-difluoro-4-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

148577-5G 148577-1G 148577-25G 148577-500MG
77631-336EA 1484.23 USD
77631-336 77636-666 77653-592 77648-710
2-(Cyclopropylmethoxy)-1,3-difluoro-4-iodobenzene ≥97%
2-(Cyclopropylmethoxy)-1,3-difluoro-4-iodobenzene

Precaución: For Research Use Only

Formula: 310.079 g/mol
MW: C₁₀H₉F₂IO
Storage Temperature: Ambient
MDL Number: MFCD34840173
CAS Number: 2921695-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR