2-Formyl-3-methoxybenzoic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
14760-1G
14760-5G
14760-500MG
77634-044EA
1126
USD
77634-044
77657-478
77648-704
2-Formyl-3-methoxybenzoic acid ≥95%
2-Formyl-3-methoxybenzoic acid
Precaución: For Research Use Only
Formula:
C₉H₈O₄ Storage Temperature: Ambient |
MDL Number:
MFCD18452452 CAS Number: 60407-04-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...