Ethyl 2-(5-chloro-2-fluoro-4-methylphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

147510-5G 147510-1G 147510-500MG
77631-328EA 1484.23 USD
77631-328 77640-676 77648-696
Ethyl 2-(5-chloro-2-fluoro-4-methylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(5-chloro-2-fluoro-4-methylphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 244.647 g/mol
MW: C₁₁H₁₀ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD34839415
CAS Number: 1859424-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR