Ethyl 2-(2-methyl-4-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

147503-5G 147503-25G 147503-1G 147503-500MG
77631-326EA 1484.23 USD
77631-326 77635-456 77636-662 77648-694
Ethyl 2-(2-methyl-4-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(2-methyl-4-(trifluoromethyl)phenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 260.209 g/mol
MW: C₁₂H₁₁F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34839409
CAS Number: 2921911-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR